Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166139.2 | NP_001159611.1 | 6 | 848 | ||
NM_153686.8 | NP_710153.2 | 6 | 848 | |||
XM_011513821.2 | XP_011512123.1 | 6 | 726 | |||
XM_011513822.2 | XP_011512124.1 | 6 | 726 | |||
XM_011513823.2 | XP_011512125.1 | 5 | 572 | |||
XM_017007961.1 | XP_016863450.1 | 5 | 2008 | |||
XM_017007962.1 | XP_016863451.1 | 5 | 650 | |||
XM_017007963.1 | XP_016863452.1 | 5 | 953 | |||
XM_017007964.1 | XP_016863453.1 | 7 | 724 | |||
XM_017007965.1 | XP_016863454.1 | 5 | 1907 | |||
GenBank mRNA | AK096181.1 | - | 7 | 709 | ||
AL700364.1 | - | 1 | 36 | |||
BC037322.1 | - | 6 | 731 | 5260413 |