Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318469.1 | NP_001305398.1 | 4 | 770 | ||
NM_001318471.1 | NP_001305400.1 | 5 | 826 | |||
NM_001318472.1 | NP_001305401.1 | 3 | 693 | |||
NM_001318473.1 | NP_001305402.1 | 3 | 643 | |||
NM_001318474.1 | NP_001305403.1 | 3 | 624 | |||
NM_001318475.1 | NP_001305404.1 | 4 | 782 | |||
NM_001318476.1 | NP_001305405.1 | 4 | 768 | |||
NM_153688.3 | NP_710155.2 | 4 | 751 | |||
XM_011522921.2 | XP_011521223.1 | 6 | 811 | |||
XM_017022983.1 | XP_016878472.1 | 4 | 938 | |||
XM_017022984.1 | XP_016878473.1 | 5 | 1065 | |||
XM_017022985.1 | XP_016878474.1 | 5 | 1070 | |||
XM_017022986.1 | XP_016878475.1 | 6 | 1158 | |||
XM_017022987.1 | XP_016878476.1 | 4 | 782 | |||
GenBank mRNA | AK094761.1 | - | 4 | 688 | ||
AK291372.1 | - | 3 | 571 | |||
AK296560.1 | - | 4 | 700 | |||
AK308134.1 | - | 3 | 600 | |||
AL038510.1 | - | 4 | 671 | |||
AL538214.3 | - | 4 | 638 | |||
BC033774.1 | - | 1 | 114 | |||
BC064604.1 | - | 4 | 629 | |||
BC113863.1 | - | 2 | 442 | |||
BC114471.1 | - | 2 | 442 | |||
BC127184.1 | - | 2 | 442 | |||
BX647144.1 | - | 2 | 406 | |||
DA781502.1 | - | 3 | 547 |