Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100598.1 | NP_001094068.1 | 6 | 1911 | ||
NM_001100599.1 | NP_001094069.1 | 5 | 1813 | |||
NM_001288805.1 | NP_001275734.1 | 8 | 2625 | |||
NM_001288806.1 | NP_001275735.1 | 6 | 1911 | |||
NM_001288807.1 | NP_001275736.1 | 6 | 1899 | |||
NM_001288808.1 | NP_001275737.1 | 4 | 1746 | |||
NM_001288809.1 | NP_001275738.1 | 4 | 1920 | |||
NM_173831.3 | NP_776192.2 | 7 | 1985 | |||
NR_110190.1 | 5 | 1846 | ||||
NR_110191.1 | 6 | 1944 | ||||
NR_110192.1 | 5 | 1849 | ||||
NR_110193.1 | 6 | 1944 | ||||
XM_005250883.3 | XP_005250940.1 | 6 | 1932 | |||
XM_006716542.3 | XP_006716605.1 | 7 | 2001 | |||
XM_006716543.3 | XP_006716606.1 | 6 | 1906 | |||
XM_011516977.2 | XP_011515279.1 | 6 | 2090 | |||
XM_011516979.2 | XP_011515281.1 | 7 | 1951 | |||
XM_017013335.1 | XP_016868824.1 | 8 | 2108 | |||
GenBank mRNA | AK001126.1 | - | 5 | 1842 | ||
AK055198.1 | - | 3 | 2731 | |||
AK055263.1 | - | 5 | 1887 | |||
AK095138.1 | - | 8 | 2568 | |||
BC014894.1 | - | 7 | 1945 | 3910185 |