Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176814.4 | NP_789784.2 | 6 | 3451 | ||
XM_005250181.3 | XP_005250238.1 | 6 | 3525 | |||
GenBank mRNA | AK125291.1 | - | 2 | 1255 | ||
BC037348.1 | - | 2 | 2425 | |||
BC110623.1 | - | 6 | 2996 | 40036036 | ||
BC110624.1 | - | 6 | 2991 | 40036038 |