Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181844.3 | NP_862827.1 | 9 | 2911 | ||
GenBank mRNA | AK024695.1 | - | 1 | 1126 | ||
AK122893.1 | - | 9 | 2957 | |||
BC059404.1 | - | 9 | 2962 | 30342006 |