Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF876P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027481.1 | 2 | 952 | |||
GenBank mRNA | BC028359.1 | - | 2 | 947 | ||
BC046475.1 | - | 2 | 922 |
Gene Symbol: ZNF595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286052.1 | NP_001272981.1 | 3 | 1192 | ||
NM_001286053.1 | NP_001272982.1 | 2 | 1065 | |||
NM_001286054.1 | NP_001272983.1 | 5 | 1389 | |||
NM_182524.3 | NP_872330.1 | 4 | 1288 | |||
XM_011513398.2 | XP_011511700.1 | 4 | 1907 | |||
GenBank mRNA | AK056302.1 | - | 4 | 1242 | ||
AK300505.1 | - | 3 | 1182 | |||
AK301848.1 | - | 2 | 1031 | |||
BC036110.1 | - | 4 | 1252 | 5297405 | ||
BC043151.1 | - | 4 | 1257 | 5297425 | ||
BX537887.1 | - | 4 | 1240 |