Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201325.1 | NP_001188254.1 | 2 | 232 | ||
NM_001201326.1 | NP_001188255.1 | 2 | 232 | |||
NM_002614.4 | NP_002605.2 | 3 | 303 | |||
XM_011509616.2 | XP_011507918.1 | 1 | 148 | |||
XM_011509618.2 | XP_011507920.1 | 2 | 236 | |||
XM_011509619.2 | XP_011507921.2 | 1 | 148 | |||
XM_017001464.1 | XP_016856953.1 | 1 | 148 | |||
XM_017001465.1 | XP_016856954.1 | 1 | 148 | |||
XM_017001466.1 | XP_016856955.1 | 1 | 148 | |||
XM_017001467.1 | XP_016856956.1 | 2 | 339 | |||
GenBank mRNA | AF012281.1 | - | 2 | 179 | ||
AK298269.1 | - | 2 | 173 | |||
AK313100.1 | - | 2 | 184 | |||
BC006496.2 | - | 2 | 243 | 2959384 | ||
BC006518.2 | - | 2 | 243 | 2959384 | ||
BX477678.1 | - | 3 | 302 |