Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182700.5 | NP_874359.2 | 2 | 288 | ||
NM_198956.3 | NP_945194.1 | 3 | 384 | |||
GenBank mRNA | AK056857.1 | - | 2 | 285 | ||
AK290465.1 | - | 2 | 288 | |||
AK299947.1 | - | 2 | 208 | |||
AK307969.1 | - | 3 | 381 | |||
AY167047.1 | - | 2 | 285 | |||
AY167048.1 | - | 2 | 273 | |||
BC038669.1 | - | 2 | 285 | 5259444 | ||
DA492955.1 | - | 2 | 288 | |||
DA497089.1 | - | 3 | 381 |