Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182700.5 | NP_874359.2 | 2 | 2828 | ||
NM_198956.3 | NP_945194.1 | 3 | 2924 | |||
GenBank mRNA | AK056857.1 | - | 3 | 2668 | ||
BC038669.1 | - | 3 | 2710 | 5259444 |