Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287437.1 | NP_001274366.1 | 6 | 793 | ||
NM_001287439.1 | NP_001274368.1 | 6 | 947 | |||
NM_001287440.1 | NP_001274369.1 | 6 | 744 | |||
NM_001287441.1 | NP_001274370.1 | 6 | 708 | |||
NM_001287442.1 | NP_001274371.1 | 6 | 701 | |||
NM_001287443.1 | NP_001274372.1 | 6 | 705 | |||
NM_024900.4 | NP_079176.2 | 6 | 829 | |||
NM_199320.3 | NP_955352.1 | 6 | 829 | |||
XM_005263232.1 | XP_005263289.1 | 6 | 671 | |||
XM_017008626.1 | XP_016864115.1 | 6 | 786 | |||
XM_017008627.1 | XP_016864116.1 | 6 | 833 | |||
GenBank mRNA | AB058710.1 | - | 2 | 129 | ||
AF520952.1 | - | 6 | 726 | |||
AK027620.1 | - | 6 | 749 | |||
AK074986.1 | - | 1 | 56 | |||
AK127326.1 | - | 6 | 601 | |||
AK304235.1 | - | 6 | 662 | |||
AK304815.1 | - | 6 | 618 | |||
BC032376.1 | - | 6 | 663 | 5111727 | ||
DB293142.1 | - | 6 | 683 |