Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243088.1 | NP_001230017.1 | 8 | 2035 | ||
NM_001243089.1 | NP_001230018.1 | 8 | 2032 | |||
NM_021953.3 | NP_068772.2 | 9 | 2080 | |||
NM_202002.2 | NP_973731.1 | 10 | 2194 | |||
NM_202003.2 | NP_973732.1 | 8 | 2035 | |||
XM_005253676.3 | XP_005253733.1 | 9 | 2083 | |||
XM_011520930.2 | XP_011519232.1 | 9 | 2080 | |||
XM_011520931.2 | XP_011519233.1 | 8 | 2038 | |||
XM_011520934.2 | XP_011519236.1 | 10 | 2141 | |||
GenBank mRNA | AK291206.1 | - | 8 | 1935 | ||
AK313845.1 | - | 8 | 1881 | |||
AY542306.1 | - | 8 | 1848 | |||
BC006192.2 | - | 8 | 1920 | 3833837 | ||
BC006529.2 | - | 8 | 1920 | 3834244 | ||
BC012863.1 | - | 8 | 1816 | 3881055 | ||
BC035437.1 | - | 2 | 1066 | |||
BT006986.1 | - | 8 | 1797 | |||
L16783.1 | - | 1 | 173 | |||
U74612.1 | - | 10 | 2025 | |||
U74613.1 | - | 8 | 1859 | |||
U83113.1 | - | 9 | 1866 |