Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005455.4 | NP_005446.2 | 11 | 2117 | ||
NM_203350.2 | NP_976225.1 | 10 | 2042 | |||
GenBank mRNA | AB209568.1 | - | 10 | 2853 | ||
AF065391.1 | - | 10 | 1778 | |||
AK023252.1 | - | 8 | 1595 | |||
AK055983.1 | - | 2 | 1412 | |||
AK222858.1 | - | 10 | 1767 | |||
AL136945.1 | - | 11 | 1839 | |||
BC033648.1 | - | 10 | 1788 | |||
BC039814.1 | - | 11 | 1863 | 6062647 | ||
BX538141.1 | - | 2 | 2655 | |||
CR749665.1 | - | 2 | 2655 |