Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000096.3 | NP_000087.1 | 1 | 353 | ||
NR_046371.1 | 1 | 353 | ||||
XM_006713499.2 | XP_006713562.1 | 1 | 358 | |||
XM_006713500.3 | XP_006713563.1 | 1 | 358 | |||
XM_006713501.2 | XP_006713564.1 | 1 | 358 | |||
XM_011512435.1 | XP_011510737.1 | 1 | 358 | |||
XM_017005734.1 | XP_016861223.1 | 1 | 358 | |||
XM_017005735.1 | XP_016861224.1 | 1 | 358 | |||
GenBank mRNA | AK095290.1 | - | 1 | 136 | ||
AK291219.1 | - | 1 | 135 | |||
AK299272.1 | - | 1 | 139 | |||
BC070304.1 | - | 1 | 306 | |||
BC094709.1 | - | 1 | 137 | |||
BC142714.1 | - | 1 | 109 | 40148840 | ||
BC146663.1 | - | 1 | 109 | 40148048 | ||
BC146801.1 | - | 1 | 133 | |||
CA391602.1 | - | 1 | 350 | |||
D00025.1 | - | 118 | ||||
DC334592.1 | - | 1 | 353 | |||
M13699.1 | - | 1 | 100 | |||
X04135.1 | - | 1 | 118 |