Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000129.3 | NP_000120.2 | 4 | 464 | ||
GenBank mRNA | AB208852.1 | - | 4 | 416 | ||
AK304335.1 | - | 3 | 246 | |||
AK312735.1 | - | 4 | 435 | |||
BC027963.1 | - | 4 | 422 | 5222962 | ||
BP251770.1 | - | 4 | 464 | |||
M14354.1 | - | 4 | 425 | |||
M14539.1 | - | 4 | 428 |