Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000129.3 | NP_000120.2 | 15 | 3075 | ||
GenBank mRNA | AB208852.1 | - | 15 | 3027 | ||
AK130666.1 | - | 8 | 1921 | |||
BC027963.1 | - | 15 | 3033 | 5222962 | ||
M14354.1 | - | 15 | 3036 | |||
M14539.1 | - | 15 | 3039 |