Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000309.3 | NP_000300.1 | 3, 4 | 449 | ||
NM_001122764.1 | NP_001116236.1 | 3, 4 | 510 | |||
XM_005245291.3 | XP_005245348.2 | 3, 4 | 488 | |||
XM_006711403.2 | XP_006711466.2 | 3, 4 | 444 | |||
XM_006711406.2 | XP_006711469.2 | 3, 4 | 346 | |||
XM_011509664.1 | XP_011507966.1 | 3, 4 | 346 | |||
XM_011509671.1 | XP_011507973.1 | 3, 4 | 346 | |||
XM_011509682.1 | XP_011507984.1 | 3, 4 | 433 | |||
XM_017001560.1 | XP_016857049.1 | 3, 4 | 488 | |||
XM_017001562.1 | XP_016857051.1 | 3, 4 | 415 | |||
XM_017001565.1 | XP_016857054.1 | 3, 4 | 415 | |||
GenBank mRNA | BC002357.2 | - | 3, 4 | 430 | 2821983 | |
D38537.1 | - | 3, 4 | 497 | |||
U26446.1 | - | 3, 4 | 411 |