Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000340.1 | NP_000331.1 | 4 | 696 | ||
NM_001278658.1 | NP_001265587.1 | 3 | 433 | |||
XM_011513087.2 | XP_011511389.1 | 5 | 443 | |||
GenBank mRNA | AK290846.1 | - | 4 | 439 | ||
AK292741.1 | - | 4 | 439 | |||
AK298418.1 | - | 3 | 197 | |||
AK313622.1 | - | 4 | 437 | |||
J03810.1 | - | 4 | 425 |