Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000341.3 | NP_000332.2 | 3 | 733 | ||
XM_011533047.2 | XP_011531349.1 | 3 | 733 | |||
GenBank mRNA | AB033549.1 | - | 3 | 714 | ||
AK091932.1 | - | 4 | 599 | |||
AK223146.1 | - | 3 | 695 | |||
AK289636.1 | - | 3 | 711 | |||
AK309145.1 | - | 3 | 727 | |||
AK313157.1 | - | 3 | 727 | |||
BC022386.1 | - | 3 | 731 | 4556946 | ||
BC093624.1 | - | 3 | 692 | 7939469 | ||
BC093626.1 | - | 3 | 687 | 7939471 | ||
DQ023513.1 | - | 3 | 655 | |||
DQ023515.1 | - | 3 | 655 | |||
DQ023516.1 | - | 3 | 655 | |||
DQ023517.1 | - | 3 | 655 | |||
KU178335.1 | - | 3 | 655 | |||
L11696.1 | - | 3 | 698 | |||
M95548.1 | - | 3 | 699 |