Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000436.3 | NP_000427.1 | 6, 7 | 999 | ||
XM_017009494.1 | XP_016864983.1 | 6, 7 | 880 | |||
GenBank mRNA | AK299668.1 | - | 2, 3 | 187 | ||
AK312327.1 | - | 6, 7 | 695 | |||
BC009001.1 | - | 6, 7 | 723 | 4148643 | ||
CR457049.1 | - | 6, 7 | 668 | |||
EF095213.1 | - | 3, 4 | 405 | |||
U62961.1 | - | 6, 7 | 766 |