Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000446.5 | NP_000437.3 | 5 | 578 | ||
GenBank mRNA | AK314027.1 | - | 5 | 504 | ||
BC074719.2 | - | 5 | 510 | 30915169 | ||
CB122285.1 | - | 5 | 538 | |||
D84371.1 | - | 5 | 535 | |||
M63012.1 | - | 5 | 490 | |||
M63013.1 | - | 5 | 429 | |||
M63014.1 | - | 4 | 395 | |||
S64615.1 | - | 5 | 490 | |||
U53784.1 | - | 5 | 535 | |||
Z70723.1 | - | 5 | 535 |