Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000550.2 | NP_000541.1 | 8 | 2452 | ||
GenBank mRNA | BC052608.1 | - | 7 | 2363 | 6258285 | |
X51420.1 | - | 8 | 2403 |