Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000698.4 | NP_000689.1 | 3 | 509 | ||
NM_001256153.2 | NP_001243082.1 | 3 | 509 | |||
NM_001256154.2 | NP_001243083.1 | 3 | 509 | |||
NM_001320861.1 | NP_001307790.1 | 3 | 509 | |||
NM_001320862.1 | NP_001307791.1 | 3 | 547 | |||
GenBank mRNA | AK225603.1 | - | 3 | 462 | ||
AK315223.1 | - | 3 | 459 | |||
BC034464.1 | - | 3 | 448 | |||
BC040450.1 | - | 3 | 438 | |||
BC130332.1 | - | 3 | 462 | 40146363 | ||
BC132677.1 | - | 3 | 462 | 40146699 | ||
BC143985.1 | - | 3 | 462 | 9052503 | ||
HM592258.1 | - | 3 | 395 | |||
HM592259.1 | - | 3 | 395 | |||
HM592260.1 | - | 3 | 395 | |||
HM592261.1 | - | 3 | 395 | |||
J03571.1 | - | 3 | 429 | |||
J03600.1 | - | 3 | 439 |