Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000802.3 | NP_000793.1 | 5 | 859 | ||
NM_016724.2 | NP_057936.1 | 6 | 1110 | |||
NM_016725.2 | NP_057937.1 | 5 | 1044 | |||
NM_016729.2 | NP_057941.1 | 4 | 879 | |||
GenBank mRNA | AK223527.1 | - | 5 | 898 | ||
BC002947.2 | - | 5 | 991 | 3956659 | ||
J05013.1 | - | 5 | 1045 | |||
M25317.1 | - | 4 | 773 | |||
M28099.1 | - | 6 | 930 | |||
X62753.1 | - | 5 | 1264 |