Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000898.4 | NP_000889.3 | 15 | 2241 | ||
XM_005272608.3 | XP_005272665.1 | 17 | 2341 | |||
XM_017029523.1 | XP_016885012.1 | 17 | 2391 | |||
XM_017029524.1 | XP_016885013.1 | 16 | 2201 | |||
GenBank mRNA | AL133791.1 | - | 1 | 77 | ||
BC022494.1 | - | 15 | 2187 | 4800208 | ||
BM726427.1 | - | 1 | 465 | |||
CA975968.1 | - | 2 | 702 | |||
M69177.1 | - | 15 | 2139 |