Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001089.2 | NP_001080.2 | 5 | 957 | ||
GenBank mRNA | AB070929.1 | - | 2 | 287 | ||
AB210044.1 | - | 5 | 957 | |||
AK310864.1 | - | 5 | 808 | |||
BC020724.1 | - | 4 | 653 | |||
BC062779.1 | - | 4 | 629 | |||
BC140895.1 | - | 2 | 282 | 9021763 | ||
BC146866.1 | - | 2 | 282 | 9056794 | ||
BC151840.1 | - | 5 | 939 | 8860312 | ||
U78735.1 | - | 5 | 804 | |||
X97187.1 | - | 4 | 592 |