Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001091.3 | NP_001082.2 | 2 | 823 | ||
NM_001272072.1 | NP_001259001.1 | 2 | 823 | |||
XM_017011944.1 | XP_016867433.1 | 4 | 1031 | |||
XM_017011945.1 | XP_016867434.1 | 4 | 1061 | |||
XM_017011946.1 | XP_016867435.1 | 2 | 987 | |||
XM_017011947.1 | XP_016867436.1 | 2 | 801 | |||
GenBank mRNA | AK092514.1 | - | 3 | 638 | ||
AK296638.1 | - | 2 | 783 | |||
AK307278.1 | - | 3 | 508 | |||
AK314806.1 | - | 2 | 781 | |||
BC014093.2 | - | 2 | 776 | 4547491 | ||
BX648159.1 | - | 4 | 4946 | |||
M55602.1 | - | 2 | 766 | |||
U11862.1 | - | 2 | 823 | |||
U11863.1 | - | 2 | 798 |