Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACACB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093.3 | NP_001084.3 | 1 | 107 | ||
XM_005253876.4 | XP_005253933.1 | 2 | 217 | |||
XM_011538259.2 | XP_011536561.1 | 2 | 855 | |||
XM_011538263.2 | XP_011536565.1 | 2 | 855 | |||
XM_011538265.2 | XP_011536567.1 | 2 | 857 | |||
GenBank mRNA | AJ575431.2 | - | 1 | 98 | ||
AJ575592.3 | - | 1 | 98 | |||
AY382667.1 | - | 2 | 217 | |||
DQ493870.1 | - | 1 | 98 | |||
U89344.1 | - | 1 | 98 |