Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLNS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293.2 | NP_001284.1 | 2 | 307 | ||
NM_001311199.1 | NP_001298128.1 | 2 | 307 | |||
NM_001311200.1 | NP_001298129.1 | 2 | 307 | |||
NM_001311201.1 | NP_001298130.1 | 2 | 307 | |||
NM_001311202.1 | NP_001298131.1 | 2 | 307 | |||
GenBank mRNA | AF005422.1 | - | 2 | 245 | ||
AF026003.1 | - | 2 | 215 | |||
AK315259.1 | - | 2 | 301 | |||
AL530926.3 | - | 2 | 253 | |||
BC119634.1 | - | 2 | 264 | 40115653 | ||
BC119635.1 | - | 2 | 264 | 40115655 | ||
BQ643786.1 | - | 2 | 277 | |||
BT019907.1 | - | 2 | 215 | |||
BU169662.1 | - | 2 | 295 | |||
BU538016.1 | - | 2 | 273 | |||
DA004269.1 | - | 2 | 308 | |||
U17899.1 | - | 2 | 289 | |||
U53454.1 | - | 2 | 299 | |||
X91788.1 | - | 2 | 303 |