Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLNS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293.2 | NP_001284.1 | 7 | 1231 | ||
NM_001311200.1 | NP_001298129.1 | 6 | 1129 | |||
NM_001311201.1 | NP_001298130.1 | 6 | 1057 | |||
NM_001311202.1 | NP_001298131.1 | 5 | 1021 | |||
GenBank mRNA | AF005422.1 | - | 7 | 1169 | ||
U17899.1 | - | 7 | 1213 | |||
U53454.1 | - | 7 | 1223 | |||
X91788.1 | - | 7 | 1227 |
Gene Symbol: LOC107987163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018644.1 | XP_016874133.1 | 1 | 122 |