Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301347.1 | NP_001288276.1 | 2 | 182 | ||
NM_001460.4 | NP_001451.2 | 3 | 320 | |||
GenBank mRNA | AK304385.1 | - | 2 | 90 | ||
AK312877.1 | - | 3 | 226 | |||
AL833218.1 | - | 3 | 209 | |||
BC005894.1 | - | 3 | 226 | 4293031 | ||
BI766311.1 | - | 3 | 324 | |||
BT006979.1 | - | 2 | 162 | |||
DB224311.1 | - | 3 | 283 | |||
DB236882.1 | - | 3 | 283 | |||
Y09267.1 | - | 3 | 222 |