Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300868.1 | NP_001287797.1 | 14 | 2104 | ||
NM_001300869.1 | NP_001287798.1 | 13 | 1935 | |||
NM_001532.2 | NP_001523.2 | 12 | 1903 | |||
NR_125343.1 | 12 | 1863 | ||||
XM_011544974.1 | XP_011543276.1 | 13 | 2253 | |||
XM_017017630.1 | XP_016873119.1 | 11 | 2456 | |||
XM_017017632.1 | XP_016873121.1 | 14 | 2054 | |||
XM_017017635.1 | XP_016873124.1 | 11 | 2083 | |||
XM_017017637.1 | XP_016873126.1 | 11 | 2123 | |||
XM_017017638.1 | XP_016873127.1 | 9 | 1626 | |||
GenBank mRNA | AF034102.1 | - | 12 | 1911 | ||
AF401235.1 | - | 12 | 1871 | |||
BC011387.1 | - | 12 | 1650 | 3872824 | ||
X86681.1 | - | 12 | 1669 |