Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001606.4 | NP_001597.2 | 38 | 6060 | ||
NM_212533.2 | NP_997698.1 | 38 | 6052 | |||
XM_006716996.3 | XP_006717059.1 | 37 | 5994 | |||
GenBank mRNA | AB028985.2 | - | 24 | 3917 | ||
AB177854.1 | - | 25 | 3961 | |||
AF178941.1 | - | 37 | 5962 | |||
AF327657.1 | - | 37 | 5918 | |||
AK295489.1 | - | 4 | 528 | |||
BC008755.1 | - | 9 | 1207 | |||
BC144588.1 | - | 37 | 6042 | |||
DB168116.1 | - | 4 | 520 |