Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001756.3 | NP_001747.2 | 4 | 1006 | ||
GenBank mRNA | AK225591.1 | - | 5 | 916 | ||
AK290821.1 | - | 4 | 991 | |||
AK313760.1 | - | 4 | 991 | |||
BC017462.1 | - | 4 | 960 | |||
BC056259.1 | - | 4 | 981 | 5810260 | ||
BC058021.1 | - | 4 | 990 | 6714173 | ||
J02943.1 | - | 3 | 937 |