Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323565.1 | NP_001310494.1 | 6 | 1082 | ||
NM_001323566.1 | NP_001310495.1 | 5 | 1022 | |||
NM_001323567.1 | NP_001310496.1 | 5 | 618 | |||
NM_001801.2 | NP_001792.2 | 5 | 1025 | |||
NR_136618.1 | 7 | 1345 | ||||
NR_136619.1 | 6 | 1101 | ||||
NR_136620.1 | 5 | 985 | ||||
NR_136621.1 | 6 | 694 | ||||
GenBank mRNA | AK130357.1 | - | 5 | 987 | ||
BC024241.2 | - | 5 | 975 | 4797463 | ||
BG197197.1 | - | 4 | 502 | |||
BG720517.1 | - | 6 | 699 | |||
CK005322.1 | - | 4 | 464 | |||
D85777.1 | - | 5 | 943 | |||
Z31357.1 | - | 5 | 968 |