Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX4I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318786.1 | NP_001305715.1 | 5 | 611 | ||
NM_001318794.1 | NP_001305723.1 | 5 | 872 | |||
NM_001318797.1 | NP_001305726.1 | 5 | 455 | |||
NM_001861.4 | NP_001852.1 | 5 | 590 | |||
GenBank mRNA | AK027136.1 | - | 1 | 3713 | ||
AK311847.1 | - | 5 | 462 | |||
BC008704.1 | - | 5 | 448 | 2990066 | ||
BC021236.2 | - | 5 | 441 | 4309698 | ||
BC047869.1 | - | 3 | 2036 | |||
BC062437.1 | - | 5 | 469 | 6451700 | ||
BE615615.1 | - | 4 | 409 | |||
BE728651.1 | - | 5 | 452 | |||
BT019825.1 | - | 4 | 403 | |||
BU599266.1 | - | 449 | ||||
DB502762.1 | - | 5 | 455 | |||
M21575.1 | - | 5 | 437 | |||
M34600.1 | - | 4 | 403 | |||
U90915.1 | - | 5 | 473 | |||
X54802.1 | - | 5 | 465 |