Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001866.2 | NP_001857.1 | 3 | 405 | ||
GenBank mRNA | BC018386.1 | - | 3 | 388 | 3861730 | |
Z14244.1 | - | 3 | 417 |