Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001917.4 | NP_001908.3 | 4 | 524 | ||
XM_005268692.3 | XP_005268749.1 | 4 | 524 | |||
XM_011538004.2 | XP_011536306.1 | 5 | 610 | |||
XM_011538005.2 | XP_011536307.1 | 5 | 614 | |||
GenBank mRNA | AK312995.1 | - | 4 | 511 | ||
AX741655.1 | - | 4 | 514 | |||
AX741656.1 | - | 4 | 514 | |||
AX741658.1 | - | 3 | 399 | |||
AX776478.1 | - | 4 | 514 | |||
AX776479.1 | - | 4 | 514 | |||
AX776480.1 | - | 7 | 1526 | |||
AX786386.1 | - | 4 | 514 | |||
BC029057.1 | - | 4 | 494 | 5184628 | ||
BC074770.2 | - | 4 | 396 | 30915193 | ||
BX538210.1 | - | 4 | 456 | |||
DA085245.1 | - | 4 | 525 | |||
KU178015.1 | - | 3 | 371 | |||
KU178016.1 | - | 3 | 371 |