Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282692.1 | NP_001269621.1 | 8 | 1495 | ||
NM_001282693.1 | NP_001269622.1 | 9 | 1489 | |||
NM_001282694.1 | NP_001269623.1 | 8 | 1300 | |||
NM_002021.2 | NP_002012.1 | 9 | 1480 | |||
XM_005245037.3 | XP_005245094.1 | 7 | 1109 | |||
XM_005245038.3 | XP_005245095.1 | 8 | 1416 | |||
XM_006711241.3 | XP_006711304.1 | 10 | 1384 | |||
GenBank mRNA | AK097039.1 | - | 9 | 2428 | ||
AK290113.1 | - | 9 | 1466 | |||
AK296198.1 | - | 8 | 1220 | |||
AK315100.1 | - | 9 | 1439 | |||
BC047129.1 | - | 8 | 1461 | 5186727 | ||
M64082.1 | - | 9 | 1428 |