Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127204.1 | NP_001120676.1 | 3, 4 | 393 | ||
NM_001127205.1 | NP_001120677.1 | 2, 3 | 297 | |||
NM_001127206.2 | NP_001120678.1 | 2, 3 | 272 | |||
NM_001286267.1 | NP_001273196.1 | 3, 4 | 396 | |||
NM_001286268.1 | NP_001273197.1 | 2, 3 | 453 | |||
NM_001286269.1 | NP_001273198.1 | 3, 4 | 374 | |||
NM_001286270.1 | NP_001273199.1 | 4, 5 | 673 | |||
NM_002134.3 | NP_002125.3 | 2, 3 | 275 | |||
XM_011522473.1 | XP_011520775.1 | 2, 3 | 263 | |||
XM_017023196.1 | XP_016878685.1 | 3, 4 | 317 | |||
XM_017023197.1 | XP_016878686.1 | 2, 3 | 148 | |||
GenBank mRNA | AF051306.1 | - | 1, 2 | 60 | ||
AK093388.1 | - | 4, 5 | 634 | |||
AK222629.1 | - | 2, 3 | 167 | |||
AK313660.1 | - | 2, 3 | 227 | |||
BC002396.2 | - | 2, 3 | 191 | 2821444 | ||
BE561721.1 | - | 2, 3 | 309 | |||
BI225653.1 | - | 2, 3 | 274 | |||
BP329113.1 | - | 3, 4 | 275 | |||
BP368949.1 | - | 2, 3 | 297 | |||
BT019788.1 | - | 1, 2 | 86 | |||
CR456760.1 | - | 1, 2 | 86 | |||
CR541839.1 | - | 1, 2 | 86 | |||
D21243.1 | - | 1, 2 | 86 | |||
DA141643.1 | - | 3, 4 | 396 | |||
DA694137.1 | - | 3, 4 | 322 | |||
DB184091.1 | - | 2, 3 | 381 | |||
S34389.1 | - | 2, 3 | 170 |