Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166343.1 | NP_001159815.1 | 3 | 602 | ||
NM_002405.3 | NP_002396.2 | 4 | 697 | |||
NR_029413.1 | 3 | 648 | ||||
GenBank mRNA | AK292123.1 | - | 4 | 638 | ||
AK297149.1 | - | 3 | 596 | |||
AK297833.1 | - | 3 | 581 | |||
AK316555.1 | - | 3 | 589 | |||
BC094814.1 | - | 4 | 658 | 5930073 | ||
BM457814.1 | - | 4 | 652 | |||
CR456518.1 | - | 4 | 490 | |||
U94352.1 | - | 4 | 604 |