Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286431.1 | NP_001273360.1 | 7 | 1164 | ||
NM_001286432.1 | NP_001273361.1 | 6 | 1057 | |||
NM_002912.4 | NP_002903.3 | 5 | 929 | |||
XM_011536028.1 | XP_011534330.1 | 4 | 1020 | |||
XM_011536029.2 | XP_011534331.1 | 4 | 1020 | |||
XM_011536030.2 | XP_011534332.1 | 4 | 1020 | |||
XM_011536031.2 | XP_011534333.1 | 5 | 1148 | |||
XM_011536032.2 | XP_011534334.1 | 5 | 5738 | |||
XM_011536036.2 | XP_011534338.1 | 4 | 1020 | |||
XM_017011152.1 | XP_016866641.1 | 5 | 1148 | |||
XM_017011153.1 | XP_016866642.1 | 5 | 691 | |||
XM_017011154.1 | XP_016866643.1 | 5 | 576 | |||
XM_017011155.1 | XP_016866644.1 | 4 | 1020 | |||
GenBank mRNA | AF035537.1 | - | 3 | 266 | ||
AF058701.1 | - | 5 | 556 | |||
AF071798.1 | - | 4 | 794 | |||
AF078695.1 | - | 6 | 1059 | |||
AF157476.1 | - | 4 | 502 | |||
AF179428.1 | - | 5 | 630 | |||
AF179429.1 | - | 6 | 737 | |||
AK310949.1 | - | 6 | 1053 | |||
BC035823.1 | - | 4 | 728 |