Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040876.1 | NP_001035809.1 | 5, 6 | 838 | ||
NM_002940.2 | NP_002931.2 | 5, 6 | 841 | |||
GenBank mRNA | AB062293.1 | - | 5, 6 | 515 | ||
AK315198.1 | - | 5, 6 | 534 | |||
BC016283.1 | - | 5, 6 | 503 | 3909151 | ||
BT009779.1 | - | 4, 5 | 398 | |||
X76388.1 | - | 5, 6 | 515 |