Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193493.1 | NP_001180422.1 | 3 | 265 | ||
NM_003038.4 | NP_003029.2 | 3 | 899 | |||
XM_006712079.2 | XP_006712142.1 | 3 | 265 | |||
GenBank mRNA | AB026689.1 | - | 3 | 637 | ||
AK126636.1 | - | 2 | 2089 | |||
AK223486.1 | - | 3 | 861 | |||
AK295687.1 | - | 3 | 220 | |||
AK298472.1 | - | 4 | 810 | |||
AK313052.1 | - | 3 | 863 | |||
AK315986.1 | - | 4 | 821 | |||
BC026216.1 | - | 3 | 800 | 4420405 | ||
BC072423.1 | - | 3 | 859 | 6186177 | ||
DA218921.1 | - | 3 | 267 | |||
L14595.1 | - | 3 | 799 | |||
L19444.1 | - | 3 | 778 |