Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134367.3 | NP_001127839.2 | 7 | 1077 | ||
NM_003043.5 | NP_003034.2 | 7 | 1081 | |||
NR_103507.2 | 7 | 1081 | ||||
XM_006713307.2 | XP_006713370.1 | 6 | 885 | |||
XM_011534030.2 | XP_011532332.1 | 6 | 838 | |||
XM_011534031.2 | XP_011532333.1 | 8 | 1209 | |||
GenBank mRNA | AB209172.1 | - | 7 | 1007 | ||
AF346763.1 | - | 4 | 516 | |||
AK303649.1 | - | 7 | 1081 | |||
AK308417.1 | - | 6 | 683 | |||
BC137128.1 | - | 7 | 1028 | 9021125 | ||
BC137129.1 | - | 7 | 1024 | 9021126 | ||
U09220.1 | - | 5 | 756 | |||
U16120.1 | - | 8 | 1132 | |||
Z18956.1 | - | 5 | 775 |