Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003049.3 | NP_003040.1 | 5 | 1539 | ||
GenBank mRNA | BC069799.1 | - | 5 | 1487 | 7262615 | |
BC069822.1 | - | 5 | 1486 | 7262603 | ||
L21893.1 | - | 5 | 1487 |