Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002490.3 | NP_002481.2 | 2, 3 | 403 | ||
GenBank mRNA | AF047182.1 | - | 2, 3 | 267 | ||
AK291874.1 | - | 2, 3 | 281 | |||
AK316562.1 | - | 2, 3 | 277 | |||
BC002772.1 | - | 2, 3 | 264 | 3619356 | ||
CR456529.1 | - | 2, 3 | 403 | |||
CR456714.1 | - | 2, 3 | 263 | |||
CR541997.1 | - | 2, 3 | 263 |