Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003058.3 | NP_003049.2 | 10 | 1743 | ||
GenBank mRNA | AB075951.1 | - | 9 | 2916 | ||
AK290787.1 | - | 10 | 1647 | |||
BC039899.1 | - | 10 | 1624 | 5186229 | ||
X98333.1 | - | 10 | 1713 |