Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQLE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003129.3 | NP_003120.2 | 11 | 2603 | ||
XM_011517246.2 | XP_011515548.1 | 11 | 1701 | |||
GenBank mRNA | AF098865.1 | - | 11 | 1832 | ||
AK225085.1 | - | 11 | 1893 | |||
AK295302.1 | - | 11 | 1503 | |||
AK301792.1 | - | 11 | 1495 | |||
AK313384.1 | - | 11 | 1721 | |||
BC017033.1 | - | 11 | 1835 | 3846346 | ||
BX647400.1 | - | 11 | 2537 | |||
BX647605.1 | - | 11 | 2575 | |||
D78130.1 | - | 11 | 1940 |