Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003404.4 | NP_003395.1 | 7 | 1930 | ||
NM_139323.3 | NP_647539.1 | 6 | 1835 | |||
XM_017028039.1 | XP_016883528.1 | 7 | 1734 | |||
GenBank mRNA | AF107406.1 | - | 1 | 695 | ||
AL834170.1 | - | 6 | 1714 | |||
BC010352.1 | - | 1 | 582 | |||
EU794580.1 | - | 7 | 1826 |