Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098484.2 | NP_001091954.1 | 13, 14 | 1753 | ||
NM_001134742.1 | NP_001128214.1 | 13, 14 | 1753 | |||
NM_003759.3 | NP_003750.1 | 10, 11 | 1700 | |||
XM_011532390.2 | XP_011530692.1 | 8, 9 | 1239 | |||
XM_017008792.1 | XP_016864281.1 | 11, 12 | 1426 | |||
XM_017008793.1 | XP_016864282.1 | 8, 9 | 1725 | |||
GenBank mRNA | AB470072.1 | - | 12, 13 | 1643 | ||
AF004813.1 | - | 11, 12 | 1576 | |||
AF007216.1 | - | 10, 11 | 1653 | |||
AF011390.1 | - | 13, 14 | 1753 | |||
AF053753.1 | - | 12, 13 | 1636 | |||
AF053754.1 | - | 12, 13 | 1636 | |||
AF069510.1 | - | 12, 13 | 1680 | |||
AF157492.1 | - | 13, 14 | 1712 | |||
AF310248.1 | - | 12, 13 | 1636 | |||
AK226164.1 | - | 11, 12 | 1526 | |||
AK307594.1 | - | 13, 14 | 1733 | |||
AK308038.1 | - | 11, 12 | 1228 | |||
BC030977.1 | - | 10, 11 | 1700 | 4610968 | ||
EF531618.1 | - | 12, 13 | 1636 |